Cianocobalamina 96%
Bottiglia di vetro, 25 g
- :68-19-9
- :C₆₃H₈₈CoN₁₄O₁₄P
- :MFCD00151092
- :1355.38 g/mol
- :12,10152
- :320 °C
- :Ambiente
- :200-680-0
Documents
- Appearance:Red to dark red Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤12 % (20 mg, 105°C, 2 hrs) (vacuum)
- UV spectrophotometry:≥95 % (on dried substance)
- Ratio of Absorbances:3.15 to 3.45 (A361/A(547 to 559))
- Tipo di conf.:Bottiglia di vetro
- Conf:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



