Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
L(-)-Carnitine 99+%

L(-)-Carnitine 99+%

Bottiglia di vetro, 50 g
Catalog # ACRO241040500
Irritant
L(-)-Carnitine 99+%
Catalog # ACRO241040500
CAS Number:  541-15-1
L(-)-Carnitine 99+%
L(-)-Carnitine 99+%

Bottiglia di vetro, 50 g

CAS Number:  541-15-1
Price
373.00 SFr.
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
  • CAS Number:541-15-1
  • Formula:C₇H₁₅NO₃
  • MDL Number:MFCD00038747
  • Molecular Weight:161.20 g/mol
  • Melting Pt:208…212 °C (dec.)
  • Storage Temperature:Refrigerata
  • EINECS:208-768-0

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Solubility:
    (5 % in water) Clear colorless
  • Specific optical rotation:
    -29° to -32° (20°C, 589 nm) (c=1, H2O)
  • Titration with HClO4:
    ≥99.0 %

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.