2,4-Dihydroxybenzaldehyde 98%
Bottiglia di vetro, 500 g
- :95-01-2
- :C₇H₆O₃
- :MFCD00011686
- :138.12 g/mol
- :12,08324
- :220…228 °C (22 mmHg)
- :133…138 °C
- :1,448 g/cm³ (-93 °C)
- :Ambiente
- :202-383-1
Documents
- Appearance:Light beige to pink-orange Powder
- Infrared spectrum:Conforms
- Melting point:134 to 140 °C
- GC:≥97.5 %
- Tipo di conf.:Bottiglia di vetro
- Conf:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



