D-(-)-Aspartic acid 99+%
Bottiglia di vetro, 100 g
: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
- :1783-96-6
- :C₄H₇NO₄
- :MFCD00063081
- :133.1 g/mol
- :13,00845
- :>300 °C
- :Ambiente
- :217-234-6
Documents
ACRO20426
SDS
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.4 % (105°C, 3 hrs)
- Enantiomeric excess:≤0.5 % (L-enantiomer)
- Residue after ignition:≤0.1 %
- Specific optical rotation:-23.8° to -25.8° (20°C, 589 nm) (c=5, 5 N HCl)
- Titration with NaOH:≥99.0 %
- Other amino acids:≤0.3 % (TLC)
- Tipo di conf.:Bottiglia di vetro
- Conf:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



