Solfanilammide 98%
Bottiglia di plastica, 2,5 kg
: 4-Aminobenzenesulphonamide
- :63-74-1
- :H₂NC₆H₄SO₂NH₂
- :MFCD00007939
- :172.21 g/mol
- :13,09010
- :401 °C (1013 hPa)
- :163…167 °C
- :1,46 g/cm³ (20 °C)
- :Ambiente
- :200-563-4
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:163 to 167 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- HPLC:≥97.5 %
- Tipo di conf.:Bottiglia di plastica
- Conf:2,5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



