Orders LinkContactUsLinkComponent
Timolo 99%
Bottiglia di plastica, 2,5 kg
  ACRO150330025
Timolo 99%
  ACRO150330025
 :  89-83-8
Timolo 99%
Timolo 99%
Bottiglia di plastica, 2,5 kg
 :  89-83-8
679.00 SFr.
1 * 2,5 kg
 :  4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol
  •  :89-83-8
  •  :2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
  •  :MFCD00002309
  •  :150.22 g/mol
  •  :13,09476
  •  :232 °C (760 mmHg)
  •  :48…52 °C
  •  :0,96 g/cm³
  •  :102 °C
  •  :Ambiente
  •  :201-944-8
  •  :2430
  •  :8,III
  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 52 °C
  • GC:
    ≥98.5 %
  • Residue after evaporation:
    ≤0.05 %
  • Tipo di conf.:
    Bottiglia di plastica
  • Conf:
    2,5 kg