2-Nonanone 99%
Bottiglia di vetro, 500 mL
: Methyl heptyl ketone, Heptyl methyl ketone
- :821-55-6
- :C₉H₁₈O
- :MFCD00009553
- :142.24 g/mol
- :188…190 °C (760 mmHg)
- :–21 °C
- :0,826 g/cm³
- :68 °C
- :Ambiente
- :212-480-0
Documents
ACRO14878
SDS
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4200 to 1.4220 (20°C, 589 nm)
- GC:≥98.5 %
- Tipo di conf.:Bottiglia di vetro
- Conf:500 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



