Bifenile 99%
Fusto in plastica, 5 kg
: Diphenyl, Phenylbenzene, PCB No. 0, 0-PCB
- :92-52-4
- :C₁₂H₁₀
- :MFCD00003054
- :154.21 g/mol
- :13,03346
- :255 °C (760 mmHg)
- :68…72 °C
- :1,175 g/cm³ (20 °C)
- :113 °C
- :Ambiente
- :202-163-5
- :3077
- :9,III
- Appearance:White to light yellow Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:68 to 72 °C
- GC:≥98.5 %
- Tipo di conf.:Fusto in plastica
- Conf:5 kg



