Orders LinkContactUsLinkComponent
Inserts for trays H 20 series
Inserts for trays H 20 series
Nonantistatic rice paper inserts, thickness 0,05 - 0,10 mm, bianco, Paper, Yes
Codice catalogo ENTEH20-001B
Nome del fornitore:   ENTEGRIS
Inserts for trays H 20 series
Codice catalogo ENTEH20-001B
Nome del fornitore:  ENTEGRIS
Inserts for trays H 20 series
Nonantistatic rice paper inserts, thickness 0,05 - 0,10 mm, bianco, Paper, Yes
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 SET
Product Details & Documents
Nest between H 20 trays and/or covers.
  • Precut to tray size
  • Inserts provide a softer surface providing protection for more sensitive components
  • Inserts are not intended to limit part migration
  • Inserts with antistatic coatings are not recommended for use when storing devices for extended periods
Specifiche
  • Colore:
    bianco
  • Descrizione:
    Nonantistatic rice paper inserts, thickness 0,05 - 0,10 mm
  • Materiale:
    Paper
  • Seq:
    03
  • Standard:
    Yes