Inserts for trays H 20 series
Nonantistatic rice paper inserts, thickness 0,05 - 0,10 mm, bianco, Paper, Yes
Codice catalogo ENTEH20-001B
Nome del fornitore: ENTEGRIS
Vai a
Product Details & Documents
Nest between H 20 trays and/or covers.
- Precut to tray size
- Inserts provide a softer surface providing protection for more sensitive components
- Inserts are not intended to limit part migration
- Inserts with antistatic coatings are not recommended for use when storing devices for extended periods
Specifiche
- Colore:bianco
- Descrizione:Nonantistatic rice paper inserts, thickness 0,05 - 0,10 mm
- Materiale:Paper
- Seq:03
- Standard:Yes