Disposable syringes
Luer lock syringe, 10 ml
Codice catalogo NSCAS7515-10
Nome del fornitore: THERMO NATIONAL SCIENTIFIC
Vai a
Product Details & Documents
These syringes are an ideal choice for all syringe filter applications. They feature a two-part construction using PP barrels and PE plungers. This eliminates the need for rubber or synthetic plunger gaskets and requires no silicone or oil lubricant in the barrel.
- Non sterile
- Bulk packed
Specifiche
- Volume:10 ml
- Descrizione:Luer lock syringe
- Seq:03