Orders LinkContactUsLinkComponent
Cryopreservation cane
Cryo cane, for Ø 12,5 mm cryo tubes
  BARN4000217
Cryopreservation cane
  BARN4000217
Cryopreservation cane
Cryopreservation cane
Cryo cane, for Ø 12,5 mm cryo tubes
SFr. 6.80
1 * 1 ST
These cane systems made of aluminium are used to store cell, tissues or other biological samples.
  • Cost effective
  • Durable aluminum construction and vacuum insulation
  • Narrow-mouth design minimises LN2 evaporation
  • Secure locking clasp
Colour: Silver
  • Descrizione:
    Cryo cane, for Ø 12,5 mm cryo tubes
  • Seq:
    02