Cryopreservation cane
Cryo cane, for Ø 12,5 mm cryo tubes
BARN4000217
These cane systems made of aluminium are used to store cell, tissues or other biological samples.
- Cost effective
- Durable aluminum construction and vacuum insulation
- Narrow-mouth design minimises LN2 evaporation
- Secure locking clasp
Colour: Silver
- Descrizione:Cryo cane, for Ø 12,5 mm cryo tubes
- Seq:02