Orders LinkContactUsLinkComponent
Accessori
Transmitted-light illuminator with 6 V, 30 W halogen bulb
Catalog # ZEIS4155101200001
Supplier:   ZEISS
Accessori
Catalog # ZEIS4155101200001
Supplier:  ZEISS
Accessori
Accessori
Transmitted-light illuminator with 6 V, 30 W halogen bulb
Price
SFr. 164.35
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Descrizione:
    Transmitted-light illuminator with 6 V, 30 W halogen bulb
  • Seq:
    29