Accessori
Microscope stand (only) with binocular ergotube, transmitted-light illumination with 6 V, 30 W halogen bulb, metal stage insert (D = 110 mm)
Catalog # ZEIS4155101102000
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Descrizione:Microscope stand (only) with binocular ergotube, transmitted-light illumination with 6 V, 30 W halogen bulb, metal stage insert (D = 110 mm)
- Seq:12