Standard hazard warning signs
Pressurized cylinder, In PE, 200×200 mm
Codice catalogo 116-0486
Nome del fornitore: Brady
Vai a
Product Details & Documents
Warning signs must have a black pictogram on a yellow triangle with a black border. Hazard signs must be displayed to warn people to be careful or to take precaution where hazards exist.
- Laminated polyester
- Resistance to most chemical products and solvents
- Resistance against extreme temperatures
- Good weatherability and UV resistance
- Excellent adhesion to low surface energy surfaces
- Thickness: 0,1 mm
Specifiche
- Descrizione:Pressurized cylinder
- Dimensioni:200×200 mm
- Materiale:In PE
- Seq:20
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



