Orders LinkContactUsLinkComponent
Phase separating filters
Phase separating filters
108 H, 270 mm
Codice catalogo PRATA270108H
Nome del fornitore:   PRAT DUMAS
Phase separating filters
Codice catalogo PRATA270108H
Nome del fornitore:  PRAT DUMAS
Phase separating filters
Phase separating filters
108 H, 270 mm
Price
SFr. 236.00
Log in per disponibilità
1 * 100 ST
Product Details & Documents
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Informazioni per l'ordine: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
Specifiche
  • Grado:
    108 H
  • Seq:
    07
  • Taglia:
    270 mm