Orders LinkContactUsLinkComponent
Bottle racks
Bottle racks
Bottle rack, 15× 100 ml (max. 50 mm diameter)
Codice catalogo HEIP1465260001
Nome del fornitore:   MERCK LIFE SCIENCE
Bottle racks
Codice catalogo HEIP1465260001
Nome del fornitore:  MERCK LIFE SCIENCE
Bottle racks
Bottle rack, 15× 100 ml (max. 50 mm diameter)
Price
395.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Racks for media bottles, made of high quality stainless steel, electrolytic polished.
  • Autoclavable
  • Easy to clean
  • Good chemical and temperature resistance
Safe and convenient transport of bottles. Suitable for isolators and clean rooms.
Specifiche
  • Descrizione:
    Bottle rack, 15× 100 ml (max. 50 mm diameter)
  • Seq:
    01