Orders LinkContactUsLinkComponent
Spill kits, ADR
Spill kits, ADR
Small ADR chemical spill kit, 37 litres, includes 25 pads (41×51 mm), 4 socs (76×1220 mm), 1 pair gloves and 1 disposal bag
Codice catalogo 121-0152
Nome del fornitore:   Brady
Spill kits, ADR
Codice catalogo 121-0152
Nome del fornitore:  Brady
Spill kits, ADR
Spill kits, ADR
Small ADR chemical spill kit, 37 litres, includes 25 pads (41×51 mm), 4 socs (76×1220 mm), 1 pair gloves and 1 disposal bag
Price
SFr. 136.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
ADR spill kits are eco-friendly and portable and come in a durable see-through bag with shoulder strap. The small ADR chemical kit is designed for small spills of chemical and hazardous liquid. The large ADR maintenance kit is designed for small spills of industrial oil, water, solvents and coolants. The Re-Form ADR kit S is designed for small spills of industrial liquids except for highly aggressive acids and caustics.
  • Quick and easy access to the sorbents
  • Durable see-through bag makes it easy to check the spill response supplies
Specifiche
  • Descrizione:
    Small ADR chemical spill kit, 37 litres, includes 25 pads (41×51 mm), 4 socs (76×1220 mm), 1 pair gloves and 1 disposal bag
  • Seq:
    01