Drum spill kits
Lab pack, chemical, SKH-20
Codice catalogo BRDY813871
Nome del fornitore: Brady
Vai a
Product Details & Documents
The drum spill kit is designed for medium sized spills.
- Water and chemical resistant drum protects sorbents
- Protects sorbents against moisture, dirt and damage
- Drum can also be used as shipping container for used sorbents
- Easy access to products with screw top lid
Informazioni per l'ordine: Supplied with 12 pads (410×510 mm), 3 socs (Ø 7,6×3660 mm), 2 pillows (430×480 mm), 1 pair of gloves, 1 pair of goggles and 3 disposal bags.
Specifiche
- Descrizione:Lab pack, chemical, SKH-20
- Seq:01