Feeder cabinets
Feeder cabinets, front load and front dispense with 4 compartments, 500×400×2000 mm
Codice catalogo PALBFFCS-0540-SST
Nome del fornitore: Palbam
Vai a
Product Details & Documents
The feeder cabinets support the gowning operation. Made of stainless steel 304 with electropolished finish.
- Front access doors are used for reloading and for storage purposes
- The bottom cabinet can be used for storage or disposal
- Perspex dispenser slots ensure that garments/disposables can be extracted one at a time and do not spill out
Informazioni per l'ordine: Typically available from stock for direct shipment from the supplier. Delivered as shipped assembled or flat packed.
Documents
Palbam Class - Catalog 2018
Letteratura sul prodotto
Specifiche
- Descrizione:Feeder cabinets, front load and front dispense with 4 compartments
- Seq:01
- L×P×A:500×400×2000 mm