Safety Spectacles, Silium+
Anti-UV (5-3,1), anti-fog, anti-scratch, antracite, Smoke PC
Codice catalogo 111-1627
Nome del fornitore: Bollé Safety
Vai a
Product Details & Documents
Inspired from SILIUM, SILIUM + are comfortable and ultra lightweight PC safety glasses. SILIUM + is an excellent choice, combining style, ergonomics and performances for extended use.
- Adjustable bridge
- Non-slip bridge
- Straight, highly flexible ergonomic temples
- PLATINUM coating
Weight: 21 g
Certifications: EN 166 FT1, EN 170, EN 172 (smoke lens).
Documents
BOLESILPPSI_TDS_EN
Scheda dati
Specifiche
- Tipo:Anti-UV (5-3,1), anti-fog, anti-scratch
- Colore:antracite
- Seq:02
- Materiale lente:Smoke PC