Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Thromboxane B₂ ELISA Kit

Thromboxane B₂ ELISA Kit

Thromboxane B₂ ELISA kit, 96 wells, 96 Tests
Codice catalogo CAYM501020-96
Nome del fornitore:   Cayman Chemical
Thromboxane B₂ ELISA Kit
Codice catalogo CAYM501020-96
Nome del fornitore:  Cayman Chemical
Numero CAS:  
Thromboxane B₂ ELISA Kit
Thromboxane B₂ ELISA Kit

Thromboxane B₂ ELISA kit, 96 wells, 96 Tests

Numero CAS:  
Price
SFr. 742.00
Log in per disponibilità
1 * 96 Tests
Product Details & Documents

For the quantification of TXB2 in urine, serum, tissue culture supernatants, and other sample matrices.

  • Measure TXB2 in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure TXB2 levels down to 5 pg/ml
  • Incubation: 18 hours | Development: 90 to 120 minutes | Read: Colorimetric at 405 to 420 nm

Thromboxane A2 (TXA2) is produced from arachidonic acid by many cells and causes irreversible platelet aggregation and contraction of vascular and bronchial smooth muscle. Like most lipid mediators, TXA2 is not a circulating hormone. It is formed in response to local stimuli and exerts its effects within a short distance of its biosynthesis. TXA2 is rapidly hydrolyzed non-enzymatically to form TXB2, which is then quickly metabolized (t½ = 5-7 minutes) to urinary metabolites for clearance by the kidneys. Because of the transient nature of this compound it is difficult to accurately measure circulating levels in whole animal experimental models. In fact, it has been shown that plasma and urine levels of TXB2 are primarily due to ex vivo platelet activation and intra-renal production, respectively. Therefore, measurement of TXB2 metabolites, such as 11-dehydro TXB2, in urine and plasma may give more accurate estimates of in vivo TXA2 production. TXB2 measurement is better suited for samples that are unlikely to undergo extensive metabolism such as perfusates, lavage samples, tissue/cell culture, etc.

 : Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, and Plate(s).

Specifiche

Specifiche

  • Conf:
    96 Tests
  • Descrizione:
    Thromboxane B₂ ELISA kit, 96 wells
  • Sensibilità:
    80% B/B0: 5 pg/ml
  • Cat. No.:
    CAYM501020-96
  • Tipo di test:
    Competitive
  • Metodo di rilevamento:
    Colorimetric at 405 - 420 nm
  • Range di rilevamento:
    1,6 - 1000 pg/ml
  • Target protein:
    Thromboxane B2 (TXB₂)
  • Tempo di sviluppo:
    90 - 120 min
  • Durata del test:
    Multiple steps
  • Tempo di incubazione:
    18 hr
  • Tiipo del campione:
    Urine, serum, tissue culture supernatants, and other sample matrices
  • Formato:
    Pre-coated

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.