Wringers for mops
Stainless steel downpress flat mop wringer for VertiKlean® MAX™ and QuickTask™ mops, 480×190×203 mm
Codice catalogo 141-1434
Nome del fornitore: CONTEC
Vai a
Product Details & Documents
Variety of wringers for curved and flat mops.
- Made of stainless steel
- Can be used with curved, flat, and string mops
- Ideal for use with Contec’s 15, 20, and 25 L bucket systems
- Fully autoclavable
Specifiche
- Descrizione:Stainless steel downpress flat mop wringer for VertiKlean® MAX™ and QuickTask™ mops
- Dimensioni:480×190×203 mm
- Seq:03