giallo, 12 mm
Stopper for test tubes Ø 12 to 16 mm, polyethylene, with tongues.
#: 822075ZA
: VWR Chemicals
#: NUNC150260
: Thermo Fisher Scientific
#: 170774ZA
#: 444-0004
: VWR Collection
#: 391-0108