Bootie benches
Bootie bench, with 2 solid undershelves, dividers and screen for separation, 1000×473×550 mm
Codice catalogo PALBBB1047-2DS
Nome del fornitore: Palbam
Vai a
Product Details & Documents
Bootie benches are designed as a swing over bench and a storage solution for booties/shoes.
- Made of stainless steel 304L with electro-polished finish
- Available in three options according to the length required
Documents
Palbam Class - Catalog 2018
Letteratura sul prodotto
Specifiche
- Descrizione:Bootie bench, with 2 solid undershelves, dividers and screen for separation
- L×P×A:1000×473×550 mm