L-Carnosine
Bottle
Codice catalogo GENORC-681
Nome del fornitore: G-Biosciences
Vai a
Product Details & Documents
L-Carnosine is a white to slightly off-white crystalline powder that is used as a substrate for carnosinase-1 and other enzymes.
Molecular Biology Grade
Specifiche
Specifiche
- Tipo di conf.:Bottle
- Cat. No.:GENORC-681
- Protein/peptide name:L-Carnosine