L-Carnosine
Bottle
Codice catalogo GENORC-677
Nome del fornitore: G-Biosciences
Vai a
Product Details & Documents
L-Carnosine is a white to slightly off-white crystalline powder that is used as a substrate for carnosinase-1 and other enzymes.
Molecular Biology Grade
Specifiche
Specifiche
- Tipo di conf.:Bottle
- Cat. No.:GENORC-677
- Protein/peptide name:L-Carnosine
- Protein synonyms:H-beta-Ala-His-OH| N-beta-Alanyl-L-histidine| beta-alanyl‑L-histidine
- CAS No.:305-84-0