Potassium chloride standard solution
Potassium chloride standard solution, 745,5 mg/ml (0,01 M) KCl, 1412 µS/cm, 50 ml
HACH2974326
: Hach
- Conf:50 ml
- Descrizione:Potassium chloride standard solution, 745,5 mg/ml (0,01 M) KCl
- Conduttanza:1412 µS/cm