Potassium chloride standard solution
Potassium chloride standard solution, 74,55 mg/ml (0,001 M) KCl,, 146,9 µS/cm, 50 ml
Catalog # HACH2974226
Supplier: Hach
Jump to:
Product Details & Documents
For calibrations/accuracy checks on conductivity/salinity meters.
Certificates
Search by Lot NumberSpecifications
Specifications
- Conf:50 ml
- Descrizione:Potassium chloride standard solution, 74,55 mg/ml (0,001 M) KCl,
- Conduttanza:146,9 µS/cm