Orders LinkContactUsLinkComponent
Potassium chloride standard solution
Potassium chloride standard solution, 74,55 mg/ml (0,001 M) KCl,, 146,9 µS/cm, 50 ml
Catalog # HACH2974226
Supplier:   Hach
Potassium chloride standard solution
Catalog # HACH2974226
Supplier:  Hach
CAS Number:  
Potassium chloride standard solution
Potassium chloride standard solution
Potassium chloride standard solution, 74,55 mg/ml (0,001 M) KCl,, 146,9 µS/cm, 50 ml
CAS Number:  
Price
SFr. 27.10
Log in for availability
1 * 50 mL
Product Details & Documents
For calibrations/accuracy checks on conductivity/salinity meters.
Specifications
Specifications
  • Conf:
    50 ml
  • Descrizione:
    Potassium chloride standard solution, 74,55 mg/ml (0,001 M) KCl,
  • Conduttanza:
    146,9 µS/cm
Specifications