Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Empty vials, depyrogenated

Vials, depyrogenated, empty, 5,00 ml, 19,84×38,1 mm, Yes, With butyl stopper, 20 mm, Aluminium, +
  EPSCST5-20A
   Produzione a basso impatto
Empty vials, depyrogenated
  EPSCST5-20A
   Produzione a basso impatto
Empty vials, depyrogenated
Empty vials, depyrogenated

Vials, depyrogenated, empty, 5,00 ml, 19,84×38,1 mm, Yes, With butyl stopper, 20 mm, Aluminium, +

 
SFr. 637.00
   
1 * 50 ST
 

Clear depyrogenated sterile empty vials are type I borosilicate, assembled with butyl stoppers and aluminium seals. Used in the aseptic protocol.

  • Capacities ranges from 1 to 100 ml
  • Includes certificates of sterility and pyrogen test
 
  • Tappo incluso:
    Yes
  • Materiale tappo:
    Aluminium
  • Dimensione tappo:
    20 mm
  • Ø×A:
    19,84×38,1 mm
  • Sterili:
    +
  • Capacità:
    5,00 ml
  • Descrizione:
    Vials, depyrogenated, empty
  • Seq:
    10
  • Tipologia tappo:
    With butyl stopper
  • Cat. No.:
    EPSCST5-20A
  • Environmentally Preferable:
    Produzione a basso impatto
  • Certificazione ambientale:
    NAR Certificate of Retirement
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.