AdvanTip™ Pipette Tips
Low retention, 2 - 300 µl, +, 250, 300 μl pipettes, Racked, 96 tips/rack
Codice catalogo HAMI68009-22
Nome del fornitore: HAMILTON BONADUZ
Vai a
Product Details & Documents
AdvanTip™ precision pipette tips are perfect for routine, high-throughput work where an extensive range of universal tips and multiple packaging options are needed to meet laboratory requirements.
- Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
- Certified free from RNase, DNase, and pyrogens
- Extended length (only some tips): Prevent sample cross-contamination and protect pipettes from contamination when pipetting into long, narrow tubes or deep well plates
- Low retention tips (only some tips): Feature walls that maximise sample recovery; ideal for quantitative PCR, valuable samples and reagents
- Universal fit, can be used with Hamilton SoftGrip™ manual pipettes
Specifiche
- Confezione:Racked, 96 tips/rack
- Sterili:+
- Volume:2 - 300 µl
- Seq:05
- Stile punta:Low retention
- Per uso con:250, 300 μl pipettes