Jars, glass
Glass jar with screw thread, 2 l
Codice catalogo SPVAH02876LPS
Nome del fornitore: GRAVIS
Vai a
Product Details & Documents
Soda lime glass jars, suitable for storage of samples in the laboratory.
Specifiche
- Capacità:2 l
- Descrizione:Glass jar with screw thread
- Seq:03