Volumetric overflow flasks, without stopper
94 ml, Overflow flask
Codice catalogo NEUB0372-10-094
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Volumetric overflow flasks, without stopper
Codice catalogo NEUB0372-10-094
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Vai a
Product Details & Documents
These volumetric overflow flasks are made of borosilicate glass 3.3.
- Calibrated to deliver (TD, Ex)
- Cut and ground exactly at the volume-mark
- Without stopper
Specially designed for water analysis.
Specifiche
- Capacità:94 ml
- Descrizione:Overflow flask
- Seq:02