Funnels with short stem
20 mm, 208 mm
HUEN951200
These food safe HDPE funnels are acid and chemical resistant and are ideal for use in the laboratory, workshop and household purposes.
- Food-safe
- Good chemical resistance
- Good temperature resistance
- Ø esterno imbuto:208 mm
- Seq:04
- Ø esterno stelo:20 mm