Funnels with short stem
17 mm, 164 mm
HUEN950700
These food safe HDPE funnels are acid and chemical resistant and are ideal for use in the laboratory, workshop and household purposes.
- Food-safe
- Good chemical resistance
- Good temperature resistance
- Ø esterno imbuto:164 mm
- Seq:03
- Ø esterno stelo:17 mm