Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

12 mm, 100 mm
Codice catalogo HUEN950400
Nome del fornitore:   hünersdorff GmbH
Funnels with short stem
Codice catalogo HUEN950400
Nome del fornitore:  hünersdorff GmbH
Funnels with short stem
Funnels with short stem

12 mm, 100 mm

Price
Richiedi prezzo
Log in per disponibilità
1 * 12 ST
Product Details & Documents

These food safe HDPE funnels are acid and chemical resistant and are ideal for use in the laboratory, workshop and household purposes.

  • Food-safe
  • Good chemical resistance
  • Good temperature resistance
Specifiche
  • Ø esterno imbuto:
    100 mm
  • Seq:
    02
  • Ø esterno stelo:
    12 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.