Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

11 mm, 80 mm
Catalog # HUEN950300
Supplier:   hünersdorff GmbH
Funnels with short stem
Catalog # HUEN950300
Supplier:  hünersdorff GmbH
Funnels with short stem
Funnels with short stem

11 mm, 80 mm

Price
SFr. 24.20
Log in for availability
1 * 12 ST
Product Details & Documents

These food safe HDPE funnels are acid and chemical resistant and are ideal for use in the laboratory, workshop and household purposes.

  • Food-safe
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Ø esterno imbuto:
    80 mm
  • Seq:
    01
  • Ø esterno stelo:
    11 mm