Buckets
nero, 10,5 L, 286 mm, 283 mm
Codice catalogo HUEN941200
Nome del fornitore: hünersdorff GmbH
Vai a
Product Details & Documents
HDPE, translucent.
- Spouted buckets for safe handling
- Embossed scale and plastic handle
- Food-safe
For industrial and laboratory applications.
Specifiche
- Capacità:10,5 L
- Colore:nero
- Altezza:283 mm
- Seq:02
- Ø:286 mm