Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
BEARD MASK COLOR: BLUE SIZE: L DESCRIPTION: LATEXFREE ELASTIC MATERIAL: FINE MICROMESH PACKAGING: 100 PCS/BAG, 1000 PCS/CARTON 1 * 1.000 pezzi
GUNA139151
:
VWR Collection
BEARD MASK COLOR: BLUE SIZE: L DESCRIPTION: LATEXFREE ELASTIC MATERIAL: FINE MICROMESH PACKAGING: 100 PCS/BAG, 1000 PCS/CARTON 1 * 1.000 pezzi
GUNA139151
:
VWR Collection
Frequently Bought Together
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
VWR®, Pipette sierologiche, Standard Line
: VWR Collection
Guanti in nitrile, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., grado gradiente for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
: VWR Chemicals
VWR®, Guanti in Nitrile
: VWR Collection
VWR®, Salviette per il viso
: VWR Collection
Strisce reattive pH, MQuant®, Supelco®
: Merck
Acetonitrile, anidro (max. 0,003% H<sub>2</sub>O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade adatto per UPLC/UHPLC
: VWR Chemicals
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
VWR® NITRILE LIGHT, Guanti in nitrile
: VWR Collection