Orders LinkContactUsLinkComponent
Galtek® check valves
Galtek® check valves
Inlet 1/4" FNPT, outlet 1/4" FNPT, length 60,7 mm
Codice catalogo ENTECK4-4N-4N
Nome del fornitore:   ENTEGRIS
Galtek® check valves
Codice catalogo ENTECK4-4N-4N
Nome del fornitore:  ENTEGRIS
Galtek® check valves
Inlet 1/4" FNPT, outlet 1/4" FNPT, length 60,7 mm
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
  • O-ring free design reduces contamination
  • All wetted parts are molded PFA and PTFE for corrosion resistance
  • No metal parts are used that could corrode or contribute to contamination
  • Poppet design allows for a high flow through rate and maximises rinsability
Specifiche
  • Descrizione:
    Inlet 1/4" FNPT, outlet 1/4" FNPT, length 60,7 mm