Cryopreservation cane
Cryo cane, 1,2 ml - 6 ampule, holds 6 samples
Catalog # BARN4000211
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These cane systems made of aluminium are used to store cell, tissues or other biological samples.
- Cost effective
- Durable aluminum construction and vacuum insulation
- Narrow-mouth design minimises LN2 evaporation
- Secure locking clasp
Colour: Silver
Specifications
- Descrizione:Cryo cane, 1,2 ml - 6 ampule, holds 6 samples
- Seq:01