Orders LinkContactUsLinkComponent
Cryopreservation cane
Cryopreservation cane
Cryo cane, 1,2 ml - 6 ampule, holds 6 samples
Catalog # BARN4000211
Cryopreservation cane
Catalog # BARN4000211
Cryopreservation cane
Cryopreservation cane
Cryo cane, 1,2 ml - 6 ampule, holds 6 samples
Price
SFr. 25.90
Log in for availability
1 * 1 ST
Product Details & Documents
These cane systems made of aluminium are used to store cell, tissues or other biological samples.
  • Cost effective
  • Durable aluminum construction and vacuum insulation
  • Narrow-mouth design minimises LN2 evaporation
  • Secure locking clasp
Colour: Silver
Specifications
  • Descrizione:
    Cryo cane, 1,2 ml - 6 ampule, holds 6 samples
  • Seq:
    01