Hydrometers in plastic
60 - 75 %mas/°B, 0,2 %mas/°B, 250 mm
SCNL3006075
High quality polycarbonate hydrometers for %mas/°Brix measures.
- Breakage protected (no glass breakage)
- High transparency with easy-to-read scales
- High precision and reproducibility
Suitable for applications in food, pharmaceutical and cosmetic industries.
Reference temperature: 20 °C
: Available with works calibration certificate on request.
: In square plastic sleeve with individual serial number.
- Lunghezza:250 mm
- Seq:05
- Divisione:0,2 %mas/°B
- Range di misura:60 - 75 %mas/°B
- Cat. No.:SCNL3006075
- Temperatura di riferimento:20 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



