Orders LinkContactUsLinkComponent
Calibration gases
Calibration gas (60 L) cylinder, 57% LEL CH₄, 100 ppm CO, 15 PPM H2S, 17% O₂, N₂ (58 l) 24 months, 58 L
  DRAE6811130
 :   Dräger
Calibration gases
  DRAE6811130
 :  Dräger
 :  
Calibration gases
Calibration gases
Calibration gas (60 L) cylinder, 57% LEL CH₄, 100 ppm CO, 15 PPM H2S, 17% O₂, N₂ (58 l) 24 months, 58 L
 :  
SFr. 350.00
1 * 58 L
Calibration gas with nitrogen background in non refillable cylinder.
 : Requires a regulator and a delivery tubing.
  • Conf:
    58 L
  • Descrizione:
    Calibration gas (60 L) cylinder, 57% LEL CH₄, 100 ppm CO, 15 PPM H2S, 17% O₂, N₂ (58 l) 24 months