Calibration gases
Calibration gas (60 L) cylinder, 57% LEL CH₄, 100 ppm CO, 15 PPM H2S, 17% O₂, N₂ (58 l) 24 months, 58 L
DRAE6811130
: Dräger
Calibration gas with nitrogen background in non refillable cylinder.
: Requires a regulator and a delivery tubing.
- Conf:58 L
- Descrizione:Calibration gas (60 L) cylinder, 57% LEL CH₄, 100 ppm CO, 15 PPM H2S, 17% O₂, N₂ (58 l) 24 months