Wafer casettes
8 inch wafer cassette, Acciaio inossidabile
Codice catalogo PALBPCPC0210
Nome del fornitore: Palbam
Vai a
Product Details & Documents
The palbam class stainless steel wafer cassette is designed to replace standard plastic cassettes, in the semiconductor manufacturing process. The cassette has been designed with H Bar.
- Non-contaminating
- Non-deforming
- Fully conductive
- Withstands temperatures up to 400 ºC
- Designed for both manual and robotic handling
Documents
Palbam Class - Catalog 2018
Letteratura sul prodotto
Specifiche
- Descrizione:8 inch wafer cassette
- Materiale:Acciaio inossidabile
- Seq:02