Labels for BBP33 printers, type B-499
Printer labels, B-499, B33-126-499, Nylon, Acrilico permanente, bianco, BBP33, 38,1×19,1 mm, 1500/roll
Katalognr. BRDY143011
Anbieter: Brady
Direkt zu
Product Details & Documents
These nylon cloth labels are used for electrical and automation identification, inside-panel component labelling, wire and cable marking.
Dokumente
Technische Daten
- Stampante:BBP33
- Tipo:B-499
- Colore:bianco
- Descrizione:Printer labels
- Dimensioni:38,1×19,1 mm
- Materiale:Nylon
- Seq:25
- Modello:B33-126-499
- Confezione:1500/roll
- Adesivo:Acrilico permanente