Orders LinkContactUsLinkComponent
Labels for BBP33 printers, type B-499 | Avantor
Labels for BBP33 printers, type B-499
Printer labels, B-499, B33-126-499, Nylon, Acrilico permanente, bianco, BBP33, 38,1×19,1 mm, 1500/roll
Katalognr. BRDY143011
Anbieter:   Brady
Labels for BBP33 printers, type B-499
Katalognr. BRDY143011
Anbieter:  Brady
Labels for BBP33 printers, type B-499
Labels for BBP33 printers, type B-499
Printer labels, B-499, B33-126-499, Nylon, Acrilico permanente, bianco, BBP33, 38,1×19,1 mm, 1500/roll
Preis
286.00 SFr.
Einloggen für Verfügbarkeit
1 * 1.500 ST
Product Details & Documents
These nylon cloth labels are used for electrical and automation identification, inside-panel component labelling, wire and cable marking.
Technische Daten
  • Stampante:
    BBP33
  • Tipo:
    B-499
  • Colore:
    bianco
  • Descrizione:
    Printer labels
  • Dimensioni:
    38,1×19,1 mm
  • Materiale:
    Nylon
  • Seq:
    25
  • Modello:
    B33-126-499
  • Confezione:
    1500/roll
  • Adesivo:
    Acrilico permanente