Orders LinkContactUsLinkComponent
Labels for BMP™ 51/53 printer, type B-595
Labels for BMP™ 51/53 printer, type B-595
Label printer tape, B-595, MC-750-595-WT-BK, Vinile, Acrilico permanente, Thermal transfer, nero/bianco, BMP41, 19 mm×7,6 m, 1 label/box
Catalog # BRDY143372
Supplier:   Brady
Labels for BMP™ 51/53 printer, type B-595
Catalog # BRDY143372
Supplier:  Brady
Labels for BMP™ 51/53 printer, type B-595
Labels for BMP™ 51/53 printer, type B-595
Label printer tape, B-595, MC-750-595-WT-BK, Vinile, Acrilico permanente, Thermal transfer, nero/bianco, BMP41, 19 mm×7,6 m, 1 label/box
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
These are made from durable, low-shrink vinyl with a very aggressive adhesive. The labels adhere to irregular, curved, rough and highly textured surfaces (painted cinder blocks, uneven wood, textured plastics, paper jacketed pipes, powder coated surfaces). Also adhere to difficult low energy surface items (PVC piping, blow-moulded equipment cases, ABS plastics, recycled plastics).
Specifications
  • Stampante:
    BMP41
  • Printing method:
    Thermal transfer
  • Tipo:
    B-595
  • Colore:
    nero/bianco
  • Descrizione:
    Label printer tape
  • Dimensioni:
    19 mm×7,6 m
  • Materiale:
    Vinile
  • Seq:
    14
  • Modello:
    MC-750-595-WT-BK
  • Confezione:
    1 label/box
  • Adesivo:
    Acrilico permanente