Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Labels for BBP33 printers, type B-481

Labels for BBP33 printers, type B-481

Printer labels, StainerBondz™, B-481, B33-19-432, Poliestere, Acrilico permanente, bianco, BBP33, 19,0×22,9 mm
Codice catalogo BRDY143055
Nome del fornitore:   Brady
Labels for BBP33 printers, type B-481
Codice catalogo BRDY143055
Nome del fornitore:  Brady
Labels for BBP33 printers, type B-481
Labels for BBP33 printers, type B-481

Printer labels, StainerBondz™, B-481, B33-19-432, Poliestere, Acrilico permanente, bianco, BBP33, 19,0×22,9 mm

Price
SFr. 297.00
Log in per disponibilità
1 * 2.000 ST
Product Details & Documents

StainerBondz™ super chemical resistant polyester (B-481) is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.

  • Performs well in the commonly used hematoxilin and eosin slide staining process
  • Excellent resistance to a wide variety of chemicals, oils and water
  • Service temperature of –80 to 130 °C

StainerBondz™ B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers.

Specifiche
  • Stampante:
    BBP33
  • Tipo:
    B-481
  • Colore:
    bianco
  • Descrizione:
    Printer labels, StainerBondz™
  • Dimensioni:
    19,0×22,9 mm
  • Materiale:
    Poliestere
  • Seq:
    02
  • Modello:
    B33-19-432
  • Adesivo:
    Acrilico permanente