Orders LinkContactUsLinkComponent
Labels for BBP3x printers, type B-593, rectangular style raised profile
Raised profile push button labels, gloss finish, B-593, B30EP-171-593-SL, Poliestere, Acrilico permanente, argento, BBP30, 26,9×12,5 mm, 500 labels/box
  BRDY118019
 :   Brady
Labels for BBP3x printers, type B-593, rectangular style raised profile
  BRDY118019
 :  Brady
Labels for BBP3x printers, type B-593, rectangular style raised profile
Labels for BBP3x printers, type B-593, rectangular style raised profile
Raised profile push button labels, gloss finish, B-593, B30EP-171-593-SL, Poliestere, Acrilico permanente, argento, BBP30, 26,9×12,5 mm, 500 labels/box
SFr. 250.00
1 * 500 ST
Raised panel adhesive-taped polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches and internal connection points.
  • Can be used as rating and serial plates using alphanumerics that require nameplate quality
  • Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
  • Service temperature of −20 to 100 °C
 : UL approved.
  • Stampante:
    BBP30
  • Tipo:
    B-593
  • Colore:
    argento
  • Descrizione:
    Raised profile push button labels, gloss finish
  • Dimensioni:
    26,9×12,5 mm
  • Materiale:
    Poliestere
  • Seq:
    05
  • Modello:
    B30EP-171-593-SL
  • Confezione:
    500 labels/box
  • Adesivo:
    Acrilico permanente
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..