Orders LinkContactUsLinkComponent
Labels for BBP3x Printers, B-569
Labels for BBP3x Printers, B-569
High performance label printer tape, B-569, B30C-4000-569-BK, Poliestere, Acrilico permanente, Thermal transfer, nero, BBP31, 101 mm×30,4 m, 1 roll/box
Catalog # BRDY117929
Supplier:   Brady
Labels for BBP3x Printers, B-569
Catalog # BRDY117929
Supplier:  Brady
Labels for BBP3x Printers, B-569
Labels for BBP3x Printers, B-569
High performance label printer tape, B-569, B30C-4000-569-BK, Poliestere, Acrilico permanente, Thermal transfer, nero, BBP31, 101 mm×30,4 m, 1 roll/box
Price
SFr. 628.00
Log in for availability
1 * 1 ROU
Product Details & Documents
This high performance polyester labeling tape is used for labeling stainless steel pipes and surfaces. Applies best to very smooth surfaces and ideal for mounting to rigid panels.
Specifications
  • Stampante:
    BBP31
  • Printing method:
    Thermal transfer
  • Tipo:
    B-569
  • Colore:
    nero
  • Descrizione:
    High performance label printer tape
  • Dimensioni:
    101 mm×30,4 m
  • Materiale:
    Poliestere
  • Seq:
    41
  • Modello:
    B30C-4000-569-BK
  • Confezione:
    1 roll/box
  • Adesivo:
    Acrilico permanente