Labels for BBP3x Printers, B-569
High performance label printer tape, B-569, B30C-1125-569-PK, Poliestere, Acrilico permanente, Thermal transfer, rosa, BBP31, 29 mm×30,4 m, 1 roll/box
Catalog # BRDY117967
Supplier: Brady
Jump to:
Product Details & Documents
This high performance polyester labeling tape is used for labeling stainless steel pipes and surfaces. Applies best to very smooth surfaces and ideal for mounting to rigid panels.
Specifications
- Stampante:BBP31
- Printing method:Thermal transfer
- Tipo:B-569
- Colore:rosa
- Descrizione:High performance label printer tape
- Dimensioni:29 mm×30,4 m
- Materiale:Poliestere
- Seq:21
- Modello:B30C-1125-569-PK
- Confezione:1 roll/box
- Adesivo:Acrilico permanente