Orders LinkContactUsLinkComponent
Labels for BBP3x printers, type B-593, rectangular style raised profile | Avantor
Labels for BBP3x printers, type B-593, rectangular style raised profile
Raised profile push button labels, gloss finish, B-593, B30EP-176-593-GN, Poliestere, Acrilico permanente, verde, BBP30, 76,2×25,4 mm, 300 labels/box
Catalog # BRDY118045
Supplier:   Brady
Labels for BBP3x printers, type B-593, rectangular style raised profile
Catalog # BRDY118045
Supplier:  Brady
Labels for BBP3x printers, type B-593, rectangular style raised profile
Labels for BBP3x printers, type B-593, rectangular style raised profile
Raised profile push button labels, gloss finish, B-593, B30EP-176-593-GN, Poliestere, Acrilico permanente, verde, BBP30, 76,2×25,4 mm, 300 labels/box
Price
SFr. 512.00
Log in for availability
1 * 300 ST
Product Details & Documents
Raised panel adhesive-taped polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches and internal connection points.
  • Can be used as rating and serial plates using alphanumerics that require nameplate quality
  • Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
  • Service temperature of −20 to 100 °C
Zertifizierung: UL approved.
Specifications
  • Stampante:
    BBP30
  • Tipo:
    B-593
  • Colore:
    verde
  • Descrizione:
    Raised profile push button labels, gloss finish
  • Dimensioni:
    76,2×25,4 mm
  • Materiale:
    Poliestere
  • Seq:
    25
  • Modello:
    B30EP-176-593-GN
  • Confezione:
    300 labels/box
  • Adesivo:
    Acrilico permanente
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..